missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Protein Wnt-10a Protein
A DNA sequence encoding the Homo sapiens (Human) Protein Wnt-10a, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP9917-ye-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_079492.2 | |
Protein Wnt-10a Protein | |
Research Use Only | |
WNT10A | |
Recombinant Protein | |
Yeast | |
MPRSAPNDIL DLRLPPEPVL NANTVCLTLP GLSRRQMEVC VRHPDVAASA IQGIQIAIHE CQHQFRDQRW NCSSLETRNK IPYESPIFSR GFRESAFAYA IAAAGVVHAV SNACALGKLK ACGCDASRRG DEEAFRRKLH RLQLDALQRG KGLSHGVPEH PALPTASPGL QDSWEWGGCS PDMGFGERFS KDFLDSREPH RDIHARMRLH NNRVGRQAVM ENMRRKCKCH GTSGSCQLKT CWQVTPEFRT VGALLRSRFH RATLIRPHNR NGGQLEPGPA GAPSPAPGAP GPRRRASPAD LVYFEKSPDF CEREPRLDSA GTVGRLCNKS SAGSDGCGSM CCGRGHNILR QTRSERCHCR FHWCCFVVCE ECRITEWVSV CK | |
Greater than 90% as determined by SDS-PAGE. |
80326 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |