missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Protein SSX5 Protein
A DNA sequence encoding the Homo sapiens (Human) Protein SSX5, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6734-ec-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_066295.3 | |
Protein SSX5 Protein | |
Research Use Only | |
SSX5 | |
Recombinant Protein | |
E. coli | |
MNGDDAFVRR PRVGSQIPQK MQKHPWRQVC DRGIHLVNLS PFWKVGREPA SSIKALLCGR GEARAFDDIA KYFSEKEWEK MKASEKIIYV YMKRKYEAMT KLGFKATLPP FMRNKRVADF QGNDFDNDPN RGNQVEHPQM TFGRLQGIFP KITPEKPAEE GNDSKGVPEA SGPQNNGKQL RPSGKLNTSE KVNKTSGPKR GKHAWTHRVR ERKQLVIYEE ISDPQEDDE | |
Greater than 80% as determined by SDS-PAGE. |
6758 | |
10 μg | |
Not determined. | |
Full Length of BC016640 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |