missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Protein RMD5 homolog A Protein
A DNA sequence encoding the Homo sapiens (Human) Protein RMD5 homolog A, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8143-ec-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_073617.1 | |
Protein RMD5 homolog A Protein | |
Research Use Only | |
RMND5A | |
Recombinant Protein | |
E. coli | |
MDQCVTVERE LEKVLHKFSG YGQLCERGLE ELIDYTGGLK HEILQSHGQD AELSGTLSLV LTQCCKRIKD TVQKLASDHK DIHSSVSRVG KAIDKNFDSD ISSVGIDGCW QADSQRLLNE VMVEHFFRQG MLDVAEELCQ ESGLSVDPSQ KEPFVELNRI LEALKVRVLR PALEWAVSNR EMLIAQNSSL EFKLHRLYFI SLLMGGTTNQ REALQYAKNF QPFALNHQKD IQVLMGSLVY LRQGIENSPY VHLLDANQWA DICDIFTRDA CALLGLSVES PLSVSFSAGC VALPALINIK AVIEQRQCTG VWNQKDELPI EVDLGKKCWY HSIFACPILR QQTTDNNPPM KLVCGHIISR DALNKMFNGS KLKCPYCPME QSPGD | |
Greater than 90% as determined by SDS-PAGE. |
64795 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |