missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Protein phosphatase 1B Protein

A DNA sequence encoding the Homo sapiens (Human) Protein phosphatase 1B, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP6520-ec-10ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16053880

Cantidad Precio
1 160.00€ / 10 microgramos
10 148.99€ / 10 microgramos
Fecha estimada de envío
11-09-2020
Añadir a la cesta


Especificaciones

Especificaciones

NP_001028729.1
Protein phosphatase 1B Protein
10 μg
Not determined.
Full Length of Isoform Beta-X
Human
His-SUMO
Tris-based buffer, 50% glycerol
5495
Greater than 90% as determined by SDS-PAGE.
Research Use Only
PPM1B
Recombinant Protein
E. coli
SIVLVCFSNA PKVSDEAVKK DSELDKHLES RVEEIMEKSG EEGMPDLAHV MRILSAENIP NLPPGGGLAG KRNVIEAVYS RLNPHRESDG ASDEAEESGS QGKLVEALRQ MRINHRGNYR QLLEEMLTSY RLAKVEGEES PAEPAATATS SNSDAGNPVT MQESHTESES GLAELDSSNE DAGTKMSGEK I
Videos
Documentos