missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Protein-lysine 6-oxidase Protein
A DNA sequence encoding the Homo sapiens (Human) Protein-lysine 6-oxidase, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8882-ec-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_002308.2 | |
Protein-lysine 6-oxidase Protein | |
Research Use Only | |
Lox | |
Recombinant Protein | |
E. coli | |
PYKYSDDNPY YNYYDTYERP RPGGRYRPGY GTGYFQYGLP DLVADPYYIQ ASTYVQKMSM YNLRCAAEEN CLASTAYRAD VRDYDHRVLL RFPQRVKNQG TSDFLPSRPR YSWEWHSCHQ HYHSMDEFSH YDLLDANTQR RVAEGHKASF CLEDTSCDYG YHRRFACTAH TQGLSPGCYD TYGADIDCQW IDITDVKPGN YILKVSVNPS YLVPESDYTN NVVRCDIRYT GHHAYASGCT ISPY | |
Greater than 90% as determined by SDS-PAGE. |
4015 | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |