missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Protein ERGIC-53-like Protein

A DNA sequence encoding the Homo sapiens (Human) Protein ERGIC-53-like, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP8055-ec-200ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16052401

  • 675.00€ / 200 microgramos


Fecha estimada de envío
24-02-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_068591.2
Protein ERGIC-53-like Protein
200 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
79748
Greater than 90% as determined by SDS-PAGE.
Research Use Only
LMAN1L
Recombinant Protein
E. coli
GCPPLRRFEY KLSFKGPRLA LPGAGIPFWS HHGDAILGLE EVRLTPSMRN RSGAVWSRAS VPFSAWEVEV QMRVTGLGRR GAQGMAVWYT RGRGHVGSVL GGLASWDGIG IFFDSPAEDT QDSPAIRVLA SDGHIPSEQP GDGASQGLGS CHWDFRNRPH PFRARITYWG QRLRMSLNSG LTPSDPGEFC VDVGPLLLVP GGFFGVSAAT GTLADDHDVL SFLTFSLSEP SPEVPPQPFL EMQQLRLARQ LEGLWARLGL GTREDVTPKS DSEAQGEGER LFDLEETLGR HRRILQALRG LSKQLAQAER QWKKQLGPPG QARPDGGWAL DASCQIPSTP GRGGHLSMSL NKDSAKVGAL LHGQWTLLQA LQEMRDAAVR MAAEAQVSYL PVGIEHHFLE LDHILGLLQE ELRGPAKAAA KAPRPPGQPP RASSCLQ
Videos
Documentos