missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Programmed cell death protein 2-like Protein

A DNA sequence encoding the Homo sapiens (Human) Programmed cell death protein 2-like, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP8106-ec-50ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16052691

  • 352.00€ / 50 microgramos


Fecha estimada de envío
26-02-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_115722.1
Programmed cell death protein 2-like Protein
50 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
84306
Greater than 80% as determined by SDS-PAGE.
Research Use Only
PDCD2L
Recombinant Protein
E. coli
AAVLKPVLLG LRDAPVHGSP TGPGAWTASK LGGIPDALPT VAAPRPVCQR CGQPLALVVQ VYCPLEGSPF HRLLHVFACA CPGCSTGGAR SWKVFRSQCL QVPEREAQDA QKQGNSLAAE DWCEGADDWG SDTEEGPSPQ FTLDFGNDAS SAKDVDWTAR LQDLRLQDAV LGAAHPVPPG LPLFLPYYIC VADEDDYRDF VNLDHAHSLL RDYQQREGIA MDQLLSQSLP NDGDEKYEKT IIKSGDQTFY KFMKRIAACQ EQILRYSWSG EPLFLTCPTS EVTELPACSQ CGGQRIFEFQ LMPALVSMLK SANLGLSVEF GTILVYTCEK SCWPPNHQTP MEEFCIIQED PDELLFK
Videos
Documentos