missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Probable dimethyladenosine transferase Protein
A DNA sequence encoding the Homo sapiens (Human) Probable dimethyladenosine transferase, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8253-ec-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001335005.1 | |
Probable dimethyladenosine transferase Protein | |
Research Use Only | |
DIMT1 | |
Recombinant Protein | |
E. coli | |
MPKVKSGAIG RRRGRQEQRR ELKSAGGLMF NTGIGQHILK NPLIINSIID KAALRPTDVV LEVGPGTGNM TVKLLEKAKK VVACELDPRL VAELHKRVQG TPVASKLQVL VGDVLKTDLP FFDTCVANLP YQISSPFVFK LLLHRPFFRC AILMFQREFA LRLVAKPGDK LYCRLSINTQ LLARVDHLMK VGKNNFRPPP KVESSVVRIE PKNPPPPINF QEWDGLVRIT FVRKNKTLSA AFKSSAVQQL LEKNYRIHCS VHNIIIPEDF SIADKIQQIL TSTGFSDKRA RSMDIDDFIR LLHGFNAEGI HFS | |
Greater than 90% as determined by SDS-PAGE. |
27292 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |