missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PRKAB2 Protein
A DNA sequence encoding the Homo sapiens (Human) PRKAB2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7495-ec-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_005390.1 | |
PRKAB2 Protein | |
Research Use Only | |
PRKAB2 | |
Recombinant Protein | |
E. coli | |
MGNTTSDRVS GERHGAKAAR SEGAGGHAPG KEHKIMVGST DDPSVFSLPD SKLPGDKEFV SWQQDLEDSV KPTQQARPTV IRWSEGGKEV FISGSFNNWS TKIPLIKSHN DFVAILDLPE GEHQYKFFVD GQWVHDPSEP VVTSQLGTIN NLIHVKKSDF EVFDALKLDS MESSETSCRD LSSSPPGPYG QEMYAFRSEE RFKSPPILPP HLLQVILNKD TNISCDPALL PEPNHVMLNH LYALSIKDSV MVLSATHRYK KKYVTTLLYK PI | |
Greater than 80% as determined by SDS-PAGE. |
5565 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |