missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Pre-mRNA-splicing factor SPF27 Protein
A DNA sequence encoding the Homo sapiens (Human) Pre-mRNA-splicing factor SPF27, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5706-ec-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_005863.1 | |
Pre-mRNA-splicing factor SPF27 Protein | |
Research Use Only | |
BCAS2 | |
Recombinant Protein | |
E. coli | |
AGTGLVAGEV VVDALPYFDQ GYEAPGVREA AAALVEEETR RYRPTKNYLS YLTAPDYSAF ETDIMRNEFE RLAARQPIEL LSMKRYELPA PSSGQKNDIT AWQECVNNSM AQLEHQAVRI ENLELMSQHG CNAWKVYNEN LVHMIEHAQK ELQKLRKHIQ DLNWQRKNMQ LTAGSKLREM ESNWVSLVSK NYEIERTIVQ LENEIYQIKQ QHGEANKENI RQDF | |
Greater than 80% as determined by SDS-PAGE. |
10286 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |