missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PRDX6 Protein
A cDNA sequence encoding the PRDX6 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10842-20ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
PRDX6 Protein | |
Research Use Only | |
The specific activity was found to be approximately 95-120 pmole/min/microg. The enzymatic activity was confirmed by measuring the remaining peroxide after incubation of PRDX6 and peroxide for 20 min at room temperature. Specific activity is defined as the amount of hydroperoxide that 1ug of enzyme can reduce at 25C for 1 minute. | |
Human | |
His | |
The Peroxiredoxin-6 solution contains 20mM Tris-HCl buffer (pH 8.0) and 20% Glycerol. |
20 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MPGGLLLGDV APNFEANTTV GRIRFHDFLG DSWGILFSHP RDFTPVCTTE LGRAAKLAPE FAKRNVKLIA LSIDSVEDHL AWSKDINAYN CEEPTEKLPF PIIDDRNREL AILLGMLDPA EKDEKGMPVT ARVVFVFGPD KKLKLSILYP ATTGRNFDEI LRVVISLQLT AEKRVATPVD WKDGDSVMVL PTIPEEEAKK LFPKGVFTKE LPSGKKYLRY TPQP | |
Greater than 95.0% as determined by SDS-PAGE. |