missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PPP2R1A Protein
A cDNA sequence encoding the PPP2R1A was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13127-20ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
PPP2R1A Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMAAADGD DSLYPIAVLI DELRNEDVQL RLNSIKKLST IALALGVERT RSELLPFLTD TIYDEDEVLL ALAEQLGTFT TLVGGPEYVH CLLPPLESLA TVEETVVRDK AVESLRAISH EHSPSDLEAH FVPLVKRLAG GDWFTSRTSA CGLFSVCYPR VSSAVKAELR QYFRNLCSDD TPMVRRAAAS KLGEFAKVLE LDNVKSEIIP MFSNLASDEQ DSVRLLAVEA CVNIAQLLPQ EDLEALVMPT LRQAAEDKSW RVRYMVADKF TELQKAVGPE ITKTDLVPAF QNLMKDCEAE VRAAASHKVK EFCENLSADC RENVIMSQIL PCIKELVSDANQHVKSALAS VIMGLSPILG KDNTIEHLLP LFLAQLKDEC PEVRLNIISN LDCVNEVIGI RQLSQSLLPA IVELAEDAKW RVRLAIIEYM PLLAGQLGVE FFDEKLNSLC MAWLVDHVYA IREAATSNLK KLVEKFGKEW AHATIIPKVL AMSGDPNYLH RMTTLFCINV LSEVCGQDIT TKHMLPTVLR MAGDPVANVR FNVAKSLQKI GPILDNSTLQ SEVKPILEKL TQDQDVDVKY FAQEALTVLS LA | |
Greater than 90.0% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
PPP2R1A protein solution (1 mg/ml) containing Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |