missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PPP1R1A Protein
A cDNA sequence encoding the PPP1R1A was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13124-1mg
Detalles adicionales : Peso : 0.01000kg
Specifications
PPP1R1A Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MEQDNSPRKI QFTVPLLEPH LDPEAAEQIR RRRPTPATLV LTSDQSSPEI DEDRIPNPHL KSTLAMSPRQ RKKMTRITPT MKELQMMVEH HLGQQQQGEE PEGAAESTGT QESRPPGIPD TEVESRLGTS GTAKKTAECI PKTHERGSKE PSTKEPSTHI PPLDSKGANS VLEHHHHHH | |
Greater than 90.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The PPP1R1A solution contains 50mM Tris pH 8.0, 0.1mM PMSF, 1mM EDTA, 1mM DTT and 10% glycerol. |