missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PPA1 Protein

A cDNA sequence encoding the PPA1 was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP13113-5ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15922549

  • 103.00€ / 5 microgramos


Fecha estimada de envío
20-08-2019
Añadir a la cesta

Especificaciones

Especificaciones

PPA1 Protein
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The PPA1 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 200mM NaCl, 2mM DTT and 20% glycerol.
Greater than 95% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSHMSGFST EERAAPFSLE YRVFLKNEKG QYISPFHDIP IYADKDVFHM VVEVPRWSNA KMEIATKDPL NPIKQDVKKG KLRYVANLFP YKGYIWNYGA IPQTWEDPGH NDKHTGCCGD NDPIDVCEIG SKVCARGEII GVKVLGILAM IDEGETDWKV IAINVDDPDA ANYNDINDVK RLKPGYLEAT VDWFRRYKVP DGKPENEFAF NAEFKDKDFA IDIIKSTHDH WKALVTKKTN GKGISCMNTT LSESPFKCDP DAARAIVDAL PPPCESACTV PTDVDKWFHH QKN
Videos
Documentos