missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human POLE3 Protein
A cDNA sequence encoding the POLE3 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13093-5ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
POLE3 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMAERPED LNLPNAVITR IIKEALPDGV NISKEARSAI SRAASVFVLY ATSCANNFAM KGKRKTLNAS DVLSAMEEME FQRFVTPLKE ALEAYRREQK GKKEASEQKK KDKDKKTDSE EQDKSRDEDN DEDEERLEEE EQNEEEEVDN | |
Greater than 90.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The POLE3 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 20% glycerol and 1mM DTT. |