missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PMP2 / FABP8 Protein
A DNA sequence encoding the Homo sapiens (Human) PMP2 / FABP8, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8590-ec-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_002668.1 | |
PMP2 / FABP8 Protein | |
Research Use Only | |
PMP2 | |
Recombinant Protein | |
E. coli | |
MSNKFLGTWK LVSSENFDDY MKALGVGLAT RKLGNLAKPT VIISKKGDII TIRTESTFKN TEISFKLGQE FEETTADNRK TKSIVTLQRG SLNQVQRWDG KETTIKRKLV NGKMVAECKM KGVVCTRIYE KV | |
Greater than 90% as determined by SDS-PAGE. |
5375 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |