missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Plasminogen Protein

A DNA sequence encoding the Human Plasminogen, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP10364-ec-100ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15933628

  • 352.00€ / 100 microgramos


Fecha estimada de envío
08-07-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_000292.1
Plasminogen Protein
100 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
VYLSECKTGN GKNYRGTMSK TKNGITCQKW SSTSPHRPRF SPATHPSEGL EENYCRNPDN DPQGPWCYTT DPEKRYDYCD ILECEEECMH CSGENYDGKI SKTMSGLECQ AWDSQSPHAH GYIPSKFPNK NLKKNYCRNP DRELRPWCFT TDPNKRWELC DIPRCTTPPP SSGPTYQCLK GTGENYRGNV AVTVSGHTCQ HWSAQTPHTH NRTPENFPCK NLDENYCRNP DGKRAPWCHT TNSQVRWEYC KIPSCDSSP
5340
Greater than 95% as determined by SDS-PAGE and HPLC.
Research Use Only
PLG
The specific activity determined by an assay on anti-proliferation and anti-migration using endothelial cells in vitro and anti-angiogenesis in vivo is 5.5 M 105 IU/mg.
Human
Untagged
0.2 um filtered 20 mM NaAc, pH 5.5, 4 % mannitol, lyophilized
Videos
Documentos