missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PL 12 Protein
A cDNA sequence encoding the PL 12 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13068-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
PL 12 Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
MDSTLTASEI RQRFIDFFKR NEHTYVHSSA TIPLDDPTLL FANAGMNQFK PIFLNTIDPS HPMAKLSRAA NTQKCIRAGG KHNDLDDVGK DVYHHTFFEM LGSWSFGDYF KELACKMALE LLTQEFGIPI ERLYVTYFGG DEAAGLEADL ECKQIWQNLG LDDTKILPGN MKDNFWEMGD TGPCGPCSEI HYDRIGGRDA AHLVNQDDPN VLEIWNLVFI QYNREADGIL KPLPKKSIDT GMGLERLVSV LQNKMSNYDT DLFVPYFEAI QKGTGARPYT GKVGAEDADG IDMAYRVLAD HARTITVALA DGGRPDNTGR GYVLRRILRR AVRYAHEKLN ASRGFFATLV DVVVQSLGDA FPELKKDPDM VKDIINEEEV QFLKTLSRGR RILDRKIQSL GDSKTIPGDT AWLLYDTYGF PVDLTGLIAE EKGLVVDMDG FEEERKLAQL KSQGKGAGGE DLIMLDIYAI EELRARGLEV TDDSPKYNYH LDSSGSYVFE NTVATVMALR REKMFVEEVS TGQECGVVLD KTCFYAEQGG QIYDEGYLVK VDDSSEDKTE FTVKNAQVRG GYVLHIGTIY GDLKVGDQVW LFIDEPRRRP IMSNHTATHI LNFALRSVLG EADQKGSLVA PDRLRFDFTA KGAMSTQQIK KAEEIANEMI EAAKAVYTQD CPLAAAKAIQ GLRAVFDETY PDPVRVVSIG VPVSELLDDP SGPAGSLTSV EFCGGTHLRN SSHAGAFVIV TEEAIAKGIR RIVAVTGAEA QKALRKAESL KKCLSVMEAK VKAQTAPNKD VQREIADLGE ALATAVIPQW QKDELRETLK SLKKVMDDLD RASKADVQKR VLEKTKQFID SNPNQPLVIL EMESGASAKA LNEALKLFKM HSPQTSAMLF TVDNEAGKIT CLCQVPQNAA | |
Greater than 90% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
PL-12 is supplied in16mM HEPES buffer pH 8.0, 250mM sodium chloride, and 20% glycerol. |