missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PIGF / PLGF Protein
A DNA sequence encoding the Homo sapiens (Human) PIGF / PLGF, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6485-ec-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001193941.1 | |
PIGF / PLGF Protein | |
Research Use Only | |
PGF | |
Recombinant Protein | |
E. coli | |
LPAVPPQQWA LSAGNGSSEV EVVPFQEVWG RSYCRALERL VDVVSEYPSE VEHMFSPSCV SLLRCTGCCG DENLHCVPVE TANVTMQLLK IRSGDRPSYV ELTFSQHVRC ECRPLREKMK PERCGDAVPR R | |
Greater than 90% as determined by SDS-PAGE. |
5228 | |
10 μg | |
Not determined. | |
Full Length of Isoform PlGF-2 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |