missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Phosphatidylserine synthase 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Phosphatidylserine synthase 1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8474-ec-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001277154.1 | |
Phosphatidylserine synthase 1 Protein | |
Research Use Only | |
PTDSS1 | |
Recombinant Protein | |
E. coli | |
MASCVGSRTL SKDDVNYKMH FRMINEQQVE DITID | |
Greater than 90% as determined by SDS-PAGE. |
9791 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |