missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PHF13 Protein

A cDNA sequence encoding the PHF13 was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP13038-1ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15922319

Cantidad Precio
1 107.00€ / 1 microgramo
10 99.50€ / 1 microgramo
Fecha estimada de envío
09-09-2020
Añadir a la cesta


Especificaciones

Especificaciones

PHF13 Protein
1 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The PHF13 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 50% glycerol, 2mM DTT, 1mM EDTA and 250mM Imidazole.
Greater than 85% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMDSDSCA AAFHPEEYSP SCKRRRTVED FNKFCTFVLA YAGYIPYPKE ELPLRSSPSP ANSTAGTIDS DGWDAGFSDI ASSVPLPVSD RCFSHLQPTL LQRAKPSNFL LDRKKTDKLK KKKKRKRRDS DAPGKEGYRG GLLKLEAADP YVETPTSPTL QDIPQAPSDP CSGWDSDTPS SGSCATVSPD QVKEIKTEGK RTIVRQGKQV VFRDEDSTGN DEDIMVDSDD DSWDLVTCFC MKPFAGRPMI ECNECHTWIH LSCAKIRKSN VPEVFVCQKC RDSKFDIRRS NRSRTGSRKL FLD
Videos
Documentos