missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PGM2 Protein

A cDNA sequence encoding the PGM2 was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP13034-1mg

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15912309

  • 3091.00€ / 1 miligramo


Fecha estimada de envío
14-08-2019
Añadir a la cesta

Especificaciones

Especificaciones

PGM2 Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
PGM2 protein solution (1 mg/ml) containing Phosphate buffered saline (pH 7.4) and 10% glycerol.
Greater than 85.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMAAPEGS GLGEDARLDQ ETAQWLRWDK NSLTLEAVKR LIAEGNKEEL RKCFGARMEF GTAGLRAAMG PGISRMNDLT IIQTTQGFCR YLEKQFSDLK QKGIVISFDA RAHPSSGGSS RRFARLAATT FISQGIPVYL FSDITPTPFV PFTVSHLKLC AGIMITASHN PKQDNGYKVY WDNGAQIISP HDKGISQAIE ENLEPWPQAW DDSLIDSSPL LHNPSASINN DYFEDLKKYC FHRSVNRETK VKFVHTSVHG VGHSFVQSAF KAFDLVPPEA VPEQKDPDPE FPTVKYPNPE EGKGVLTLSF ALADKTKARI VLANDPDADR LAVAEKQDSG EWRVFSGNEL GALLGWWLFT SWKEKNQDRS ALKDTYMLSS TVSSKILRAI ALKEGFHFEE TLTGFKWMGN RAKQLIDQGK TVLFAFEEAI GYMCCPFVLD KDGVSAAVIS AELASFLATK NLSLSQQLKA IYVEYGYHIT KASYFICHDQ ETIKKLFENL RNYDGKNNYP KACGKFEISA IRDLTTGYDD SQPDKKAVLP TSKSSQMITF TFANGGVATM RTSGTEPKIK YYAELCAPPG NSDPEQLKKE LNELVSAIEE HFFQPQKYNL QPKAD
Videos
Documentos