missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PGAM1 Protein

A cDNA sequence encoding the PGAM1 was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP10812-2ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15925248

  • 103.00€ / 2 microgramos


Fecha estimada de envío
25-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

PGAM1 Protein
2 μg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MAAYKLVLIR HGESAWNLEN RFSGWYDADL SPAGHEEAKR GGQALRDAGY EFDICFTSVQ KRAIRTLWTV LDAIDQMWLP VVRTWRLNER HYGGLTGLNK AETAAKHGEA QVKIWRRSYD VPPPPMEPDH PFYSNISKDR RYADLTEDQL PSCESLKDTI ARALPFWNEE IVPQIKEGKR VLIAAHGNSL RGIVKHLEGL SEEAIMELNL PTGIPIVYEL DKNLKPIKPM QFLGDEETVR KAMEAVAAQG KAKK
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Specific activity is >300 units/mg, in which One unit will convert 1.0 umole of 3-phosphoglycerate to 2-phosphoglcerate per minute at pH 7.6 at 37C.
Human
His
The PGAM1 1 mg/ml protein solution contains 20mM Tris-HCl pH 8.0, 1mM DTT, and 10% glycerol.
Videos