missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Peroxisomal multifunctional enzyme type 2 Protein
A DNA sequence encoding the Homo sapiens (Human) Peroxisomal multifunctional enzyme type 2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP9186-ye-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_000405.1 | |
Peroxisomal multifunctional enzyme type 2 Protein | |
Research Use Only | |
HSD17B4 | |
Recombinant Protein | |
Yeast | |
MGSPLRFDGR VVLVTGAGAG LGRAYALAFA ERGALVVVND LGGDFKGVGK GSLAADKVVE EIRRRGGKAV ANYDSVEEGE KVVKTALDAF GRIDVVVNNA GILRDRSFAR ISDEDWDIIH RVHLRGSFQV TRAAWEHMKK QKYGRIIMTS SASGIYGNFG QANYSAAKLG LLGLANSLAI EGRKSNIHCN TIAPNAGSRM TQTVMPEDLV EALKPEYVAP LVLWLCHESC EENGGLFEVG AGWIGKLRWE RTLGAIVRQK NHPMTPEAVK ANWKKICDFE NASKPQSIQE STGSIIEVLS KIDSEGGVSA NHTSRATSTA TSGFAGAIGQ KLPPFSYAYT ELEAIMYALG VGASIKDPKD LKFIYEGSSD FSCLPTFGVI IGQKSMMGGG LAEIPGLSIN FAKVLHGEQY LELYKPLPRA GKLKCEAVVA DVLDKGSGVV IIMDVYSYSE KELICHNQFS LFLVGSGGFG GKRTSDKVKV AVAIPNRPPD AVLTDTTSLN QAALYRLSGD WNPLHIDPNF ASLAGFDKPI LHGLCTFGFS ARRVLQQFAD NDVSRFKAIK ARFAKPVYPG QTLQTEMWKE GNRIHFQTKV QETGDIVISN AYVDLAPTSG TSAKTPSEGG KLQSTFVFEE IGRRLKDIGP EVVKKVNAVF EWHITKGGNI GAKWTIDLKS GSGKVYQGPA KGAADTTIIL SDEDFMEVVL GKLDPQKAFF SGRLKARGNI MLSQKLQMIL KDYAKL | |
Greater than 90% as determined by SDS-PAGE. |
3295 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |