missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Peroxisomal bifunctional enzyme Protein
A DNA sequence encoding the Homo sapiens (Human) Peroxisomal bifunctional enzyme, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7513-ec-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001159887.1 | |
Peroxisomal bifunctional enzyme Protein | |
Research Use Only | |
EHHADH | |
Recombinant Protein | |
E. coli | |
MAEYTRLHNA LALIRLRNPP VNAISTTLLR DIKEGLQKAV IDHTIKAIVI CGAEGKFSAG ADIRGFSAPR TFGLTLGHVV DEIQRNEKPV VAAIQGMAFG GGLELALGCH YRIAHAEAQV GLPEVTLGLL PGARGTQLLP RLTGVPAALD LITSGRRILA DEALKLGILD KVVNSDPVEE AIRFAQRVSD QPLESRRLCN KPIQSLPNMD SIFSEALLKM RRQHPGCLAQ EACVRAVQAA VQYPYEVGIK KEEELFLYLL QSGQARALQY AFFAERKANK WSTPSGASWK TASARPVSSV GVVGLGTMGR GIVISFARAR IPVIAVDSDK NQLATANKMI TSVLEKEASK MQQSGHPWSG PKPRLTSSVK ELGGVDLVIE AVFEEMSLKK QVFAELSAVC KPEAFLCTNT SALDVDEIAS STDRPHLVIG THFFSPAHVM KLLEVIPSQY SSPTTIATVM NLSKKIKKIG VVVGNCFGFV GNRMLNPYYN QAYFLLEEGS KPEEVDQVLE EFGFKMGPFR VSDLAGLDVG WKSRKGQGLT GPTLLPGTPA RKRGNRRYCP IPDVLCELGR FGQKTGKGWY QYDKPLGRIH KPDPWLSKFL SRYRKTHHIE PRTISQDEIL ERCLYSLINE AFRILGEGIA ASPEHIDVVY LHGYGWPRHK GGPMFYASTV GLPTVLEKLQ KYYRQNPDIP QLEPSDYLKK LASQGNPPLK EWQSLAGSPS SKL | |
Greater than 90% as determined by SDS-PAGE. |
1962 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |