missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PDZD11 / PDZK11 / PISP Protein

A DNA sequence encoding the Homo sapiens (Human) PDZD11 / PDZK11 / PISP, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP7680-ec-500ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16050141

  • 1180.00€ / 500 microgramos


Fecha estimada de envío
27-03-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_057568.1
PDZD11 / PDZK11 / PISP Protein
500 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
51248
Greater than 90% as determined by SDS-PAGE.
Research Use Only
PDZD11
Recombinant Protein
E. coli
MDSRIPYDDY PVVFLPAYEN PPAWIPPHER VHHPDYNNEL TQFLPRTITL KKPPGAQLGF NIRGGKASQL GIFISKVIPD SDAHRAGLQE GDQVLAVNDV DFQDIEHSKA VEILKTAREI SMRVRFFPYN YHRQKERTVH
Videos
Documentos