missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PDIA4 Protein

A cDNA sequence encoding the PDIA4 was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP10809-100ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15915238

  • 1257.00€ / 100 microgramos


Fecha de envío estimada
del proveedor 04-04-2019
Añadir a la cesta

Especificación

Especificación

9601
Greater than 90.0% as determined by SDS-PAGE.
Research Use Only
PDIA4
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MVAGAEGPDE DSSNRENAIE DEEEEEEEDD DEEEDDLEVK EENGVLVLND ANFDNFVADK DTVLLEFYAP WCGHCKQFAP EYEKIANILK DKDPPIPVAK IDATSASVLA SRFDVSGYPT IKILKKGQAV DYEGSRTQEE IVAKVREVSQ PDWTPPPEVT LVLTKENFDE VVNDADIILV EFYAPWCGHC KKLAPEYEKA AKELSKRSPP IPLAKVDATA ETDLAKRFDV SGYPTLKIFR KGRPYDYNGP REKYGIVDYM IEQSGPPSKE ILTLKQVQEF LKDGDDVIII GVFKGESDPA YQQYQDAANN LREDYKFHHT FSTEIAKFLK VSQGQLVVMQ PEKFQSKYEP RSHMMDVQGS TQDSAIKDFV LKYALPLVGH RKVSNDAKRY TRRPLVVVYY SVDFSFDYRA ATQFWRSKVL EVAKDFPEYT FAIADEEDYA GEVKDLGLSE SGEDVNAAIL DESGKKFAME PEEFDSDTLR EFVTAFKKGK LKPVIKSQPV PKNNKGPVKV VVGKTFDSIV MDPKKDVLIE FYAPWCGHCK QLEPVYNSLA KKYKGQKGLV IAKMDATAND VPSDRYKVEG FPTIYFAPSG DKKNPVKFEG GDRDLEHLSK FIEEHATKLS RTKEEL
PDIA4 Protein
100 μg
< 1.0 EU per ug protein as determined by the LAL method.
Specific activity > 10 A650/cm/min/mg. Enzymatic activity was confirmed by measuring the aggregation of insulin in the presence of DTT.
Human
His
The PDIA4 1 mg/ml protein solution contains 20mM Tris-HCl, pH 8.0, 1mM DTT, 0.1M NaCl and 10% glycerol.
Videos