missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PDE6H Protein

A cDNA sequence encoding the PDE6H was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP12991-2ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15922179

  • 103.00€ / 2 microgramos


Fecha estimada de envío
24-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

5149
Greater than 80% as determined by SDS-PAGE.
Research Use Only
PDE6H
Human
His
PDE6H protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 40% glycerol, 2mM DTT and 0.1mM PMSF.
PDE6H Protein
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMSDNTTL PAPASNQGPT TPRKGPPKFK QRQTRQFKSK PPKKGVKGFG DDIPGMEGLG TDITVICPWE AFSHLELHEL AQFGII
Videos