missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Paraneoplastic antigen Ma2 Protein

A DNA sequence encoding the Homo sapiens (Human) Paraneoplastic antigen Ma2, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP6510-ec-200ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16053810

  • 657.00€ / 200 microgramos


Fecha estimada de envío
19-09-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_009188.1
Paraneoplastic antigen Ma2 Protein
200 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
10687
Greater than 90% as determined by SDS-PAGE.
Research Use Only
PNMA2
Recombinant Protein
E. coli
ALALLEDWCR IMSVDEQKSL MVTGIPADFE EAEIQEVLQE TLKSLGRYRL LGKIFRKQEN ANAVLLELLE DTDVSAIPSE VQGKGGVWKV IFKTPNQDTE FLERLNLFLE KEGQTVSGMF RALGQEGVSP ATVPCISPEL LAHLLGQAMA HAPQPLLPMR YRKLRVFSGS AVPAPEEESF EVWLEQATEI VKEWPVTEAE KKRWLAESLR GPALDLMHIV QADNPSISVE ECLEAFKQVF GSLESRRTAQ VRYLKTYQEE GEKVSAYVLR LETLLRRAVE KRAIPRRIAD QVRLEQVMAG ATLNQMLWCR LRELKDQGPP PSFLELMKVI REEEEEEASF ENESIEEPEE RDGYGRWNHE
Videos
Documentos