missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Paraneoplastic antigen-like protein 1

A DNA sequence encoding the Homo sapiens (Human) Paraneoplastic antigen-like protein 5, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP7986-ec-50ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16052001

  • 234.00€ / 50 microgramos


Fecha estimada de envío
21-02-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_001096620.1
Paraneoplastic antigen-like protein 1
50 μg
Not determined.
Partial (see sequence information for more details).
Human
His-SUMO
Tris-based buffer, 50% glycerol
114824
Greater than 90% as determined by SDS-PAGE.
Research Use Only
PNMA5
Recombinant Protein
E. coli
TLLEDWCKGM DMDPRKALLI VGIPMECSEV EIQDTVKAGL QPLCAYRVLG RMFRREDNAK AVFIELADTV NYTTLPSHIP GKGGSWEVVV KPRNPDDEFL SRLNYFLKDE GRSMTDVARA LGCCSLPAES LDAEVMPQVR SPPLEPPKES MWYRKLKVFS GTASPSPGEE TFEDWLEQVT EIMPIWQVSE VEKRRRLLES LRGPALSIMR VLQANNDSIT VEQCLDALKQ IFGDKEDFRA SQFRFLQTSP KIGEKVSTFL LRLEPLLQKA VHKSPLSVRS TDMIRLKHLL ARVAMTPALR GKLELLDQRG CPPNFLELMK LIRDEEEWEN TEAVMKNKEK PSGRGRGASG RQARAEASVS APQATVQARS FSDSSPQTIQ GGLPPLVKRR RLLGSESTRG EDHGQATYPK AENQTPGREG PQAAGEELGN EAGAGAMSH
Videos
Documentos