missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human P63 / TP63 Protein
A DNA sequence encoding the Homo sapiens (Human) P63 / TP63, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8724-ec-10ug
Additional Details : Peso : 0.01000kg
Specifications
NP_001108450.1 | |
P63 / TP63 Protein | |
Research Use Only | |
TP63 | |
Recombinant Protein | |
E. coli | |
MNFETSRCAT LQYCPDPYIQ RFVETPAHFS WKESYYRSTM SQSTQTNEFL SPEVFQHIWD FLEQPICSVQ PIDLNFVDEP SEDGATNKIE ISMDCIRMQD SDLSDPMWPQ YTNLGLLNSM DQQIQNGSSS TSPYNTDHAQ NSVTAPSPYA QPSSTFDALS PSPAIPSNTD YPGPHSFDVS FQQSSTAKSA TWTYSTELKK LYCQIAKTCP IQIKVMTPPP QGAVIRAMPV YKKAEHVTEV VKRCPNHELS REFNEGQIAP PSHLIRVEGN SHAQYVEDPI TGRQSVLVPY EPPQVGTEFT TVLYNFMCNS SCVGGMNRRP ILIIVTLETR DGQVLGRRCF EARICACPGR DRKADEDSIR KQQVSDSTKN GDGTKRPFRQ NTHGIQMTSI KKRRSPDDEL LYLPVRGRET YEMLLKIKES LELMQYLPQH TIETYRQQQQ QQHQHLLQKQ TSIQSPSSYG NSSPPLNKMN SMNKLPSVSQ LINPQQRNAL TPTTIPDGMG ANIPMMGTHM PMAGDMNGLS PTQALPPPLS MPSTSHCTPP PPYPTDCSIV SFLARLGCSS CLDYFTTQGL TTIYQIEHYS MDDLASLKIP EQFRHAIWKG ILDHRQLHEF SSPSHLLRTP SSASTVSVGS SETRGERVID AVRFTLRQTI SFPPRDEWND FNFDMDARRN KQQRIKEEGE | |
Greater than 90% as determined by SDS-PAGE. |
8626 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |