missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human OSGEP Protein

A DNA sequence encoding the Homo sapiens (Human) OSGEP, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP8150-ec-500ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16052921

  • 1180.00€ / 500 microgramos


Fecha estimada de envío
20-02-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_060277.1
OSGEP Protein
500 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
55644
Greater than 80% as determined by SDS-PAGE.
Research Use Only
OSGEP
Recombinant Protein
E. coli
MPAVLGFEGS ANKIGVGVVR DGKVLANPRR TYVTPPGTGF LPGDTARHHR AVILDLLQEA LTESGLTSQD IDCIAYTKGP GMGAPLVSVA VVARTVAQLW NKPLVGVNHC IGHIEMGRLI TGATSPTVLY VSGGNTQVIA YSEHRYRIFG ETIDIAVGNC LDRFARVLKI SNDPSPGYNI EQMAKRGKKL VELPYTVKGM DVSFSGILSF IEDVAHRMLA TGECTPEDLC FSLQETVFAM LVEITERAMA HCGSQEALIV GGVGCNVRLQ EMMATMCQER GARLFATDER FCIDNGAMIA QAGWEMFRAG HRTPLSDSGV TQRYRTDEVE VTWRD
Videos
Documentos