missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Oncostatin M / OSM Protein

A DNA sequence encoding the Homo sapiens (Human) Oncostatin M / OSM, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP8301-ec-500ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16053881

  • 1210.00€ / 500 microgramos


Fecha estimada de envío
18-09-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_001306037.1
Oncostatin M / OSM Protein
500 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
5008
Greater than 90% as determined by SDS-PAGE.
Research Use Only
OSM
Recombinant Protein
E. coli
AAIGSCSKEY RVLLGQLQKQ TDLMQDTSRL LDPYIRIQGL DVPKLREHCR ERPGAFPSEE TLRGLGRRGF LQTLNATLGC VLHRLADLEQ RLPKAQDLER SGLNIEDLEK LQMARPNILG LRNNIYCMAQ LLDNSDTAEP TKAGRGASQP PTPTPASDAF QRKLEGCRFL HGYHRFMHSV GRVFSKWGES PNRSRR
Videos
Documentos