missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human OLFM1 Protein

A cDNA sequence encoding the OLFM1 was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP12923-1mg

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15911969

  • 6130.00€ / 1 miligramo


Fecha estimada de envío
20-12-2019
Añadir a la cesta

Especificaciones

Especificaciones

OLFM1 Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
OLFM1 protein solution (0.25 mg/ml) contains 20mM Tris-HCl buffer, (pH 7.5), 0.2M NaCl, 50% glycerol and 5mM DTT.
Greater than 90.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSLPTNPEE SWQVYSSAQD SEGRCICTVV APQQTMCSRD ARTKQLRQLL EKVQNMSQSI EVLDRRTQRD LQYVEKMENQ MKGLESKFKQ VEESHKQHLA RQFKG
Videos
Documentos