missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NUDCD2 Protein
A cDNA sequence encoding the NUDCD2 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12898-20ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NUDCD2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMSAPFEE RSGVVPCGTP WGQWYQTLEE VFIEVQVPPG TRAQDIQCGL QSRHVALSVG GREILKGKLF DSTIADEGTW TLEDRKMVRI VLTKTKRDAA NCWTSLLESE YAADPWVQDQ MQRKLTLERF QKENPGFDFS GAEISGNYTK GGPDFSNLEK | |
Greater than 95% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
NUDCD2 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 10% glycerol and 1mM DTT. |