missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human NTF4 Protein

A DNA sequence encoding the Human NTF4, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP10412-ec-250ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15913778

  • 2007.00€ / 250 microgramos


Fecha estimada de envío
16-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_006170.1
NTF4 Protein
250 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
MGVSETAPAS RRGELAVCDA VSGWVTDRRT AVDLRGREVE VLGEVPAAGG SPLRQYFFET RCKADNAEEG GPGAGGGGCR GVDRRHWVSE CKAKQSYVRA LTADAQGRVG WRWIRIDTAC VCTLLSRTGR A
4909
Greater than 97% as determined by SDS-PAGE and HPLC.
Research Use Only
NTF4
The ED50 as determined by the dose-dependent induction of choline acetyl transferase activity in rat basal forebrain primary septal cell cultures is less than 50 ng/ml, corresponding to a specific activity of > 2.0 M 104 IU/mg.
Human
Untagged
0.2 um filtered PBS, pH 5.5, lyophilized
Videos