missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NTF4 Protein
A DNA sequence encoding the Homo sapiens (Human) NTF4, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8678-ec-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_006170.1 | |
NTF4 Protein | |
Research Use Only | |
NTF4 | |
Recombinant Protein | |
E. coli | |
VSETAPASRR GELAVCDAVS GWVTDRRTAV DLRGREVEVL GEVPAAGGSP LRQYFFETRC KADNAEEGGP GAGGGGCRGV DRRHWVSECK AKQSYVRALT ADAQGRVGWR WIRIDTACVC TLLSRTGRA | |
Greater than 90% as determined by SDS-PAGE. |
4909 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |