missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NTF3 / Neurotrophin 3 Protein
Recombinant Protein
Marca: enQuireBio™ QP1246-10 ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001096124.1 | |
NTF3 / Neurotrophin 3 Protein | |
Research Use Only | |
NTF3 | |
Recombinant Protein | |
E. coli | |
MYAEHKSHRG EYSVCDSESL WVTDKSSAID IRGHQVTVLG EIKTGNSPVK QYFYETRCKE ARPVKNGCRG IDDKHWNSQC KTSQTYVRAL TSENNKLVGW RWIRIDTSCV CALSRKIGRT | |
Greater than 95% as determined by SDS-PAGE. |
4908 | |
10 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Shows bioactivity at a concentration of 20 ug/mL in the C6 cell proliferation assay. The bioactivity acceptance criteria is set at 50 units/mg. | |
Human | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |