missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human NTF3 / Neurotrophin 3 Protein

Recombinant Protein

Marca:  enQuireBio™ QP1246-1 mg

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15920489

  • 5607.00€ / 1 miligramo


Fecha estimada de envío
16-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_001096124.1
NTF3 / Neurotrophin 3 Protein
1 mg
Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Shows bioactivity at a concentration of 20 ug/mL in the C6 cell proliferation assay. The bioactivity acceptance criteria is set at 50 units/mg.
Human
Untagged
Sterile filtered (0.22 um) solution containing 0.1% TFA
4908
Greater than 95% as determined by SDS-PAGE.
Research Use Only
NTF3
Recombinant Protein
E. coli
MYAEHKSHRG EYSVCDSESL WVTDKSSAID IRGHQVTVLG EIKTGNSPVK QYFYETRCKE ARPVKNGCRG IDDKHWNSQC KTSQTYVRAL TSENNKLVGW RWIRIDTSCV CALSRKIGRT
Videos