missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NRG-3 / Neuregulin-3 Protein
A DNA sequence encoding the Homo sapiens (Human) NRG-3 / Neuregulin-3, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6437-ec-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001010848.2 | |
NRG-3 / Neuregulin-3 Protein | |
Research Use Only | |
NRG3 | |
Recombinant Protein | |
E. coli | |
MSEGAAAASP PGAASAAAAS AEEGTAAAAA AAAAGGGPDG GGEGAAEPPR ELRCSDCIVW NRQQTWLCVV PLFIGFIGLG LSLMLLKWIV VGSVKEYVPT DLVDSKGMGQ DPFFLSKPSS FPKAMETTTT TTSTTSPATP SAGGAASSRT PNRISTRLTT ITRAPTRFPG HRVPIRASPR STTARNTAAP ATVPSTTAPF FSSSTLGSRP PVPGTPSTQA MPSWPTAAYA TSSYLHDSTP SWTLSPFQDA ASSSSSSSSS ATTTTPETST SPKFHTTTYS TERSEHFKPC RDKDLAYCLN DGECFVIETL TGSHKHCRCK EGYQGVRCDQ FLPKTDSILS DPTDHLGIEF MESEEVYQRQ | |
Greater than 90% as determined by SDS-PAGE. |
10718 | |
10 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |