missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NPPC Protein
A cDNA sequence encoding the NPPC was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12874-20ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
4880 | |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSKPGAPPK VPRTPPAEEL AEPQAAGGGQ KKGDKAPGGG GANLKGDRSR LLRDLRVDTK SRAAWARLLQ EHPNARKYKG ANKKGLSKGC FGLKLDRIGS MSGLGC | |
Greater than 85% as determined by SDS-PAGE. |
NPPC Protein | |
Research Use Only | |
NPPC | |
Human | |
His | |
The NPPC solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl and 10% glycerol. |