missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human NMNAT2 Protein

A cDNA sequence encoding the NMNAT2 was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP10793-0.1mg

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15935188

  • 1372.00€ / 100 microgramos


Fecha estimada de envío
21-10-2019
Añadir a la cesta

Especificaciones

Especificaciones

23057
Greater than 85.0% as determined by SDS-PAGE.
Research Use Only
NMNAT2
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MTETTKTHVI LLACGSFNPI TKGHIQMFER ARDYLHKTGR FIVIGGIVSP VHDSYGKQGL VSSRHRLIMC QLAVQNSDWI RVDPWECYQD TWQTTCSVLE HHRDLMKRVT GCILSNVNTP SMTPVIGQPQ NETPQPIYQN SNVATKPTAA KILGKVGESL SRICCVRPPV ERFTFVDENA NLGTVMRYEE IELRILLLCG SDLLESFCIP GLWNEADMEV IVGDFGIVVV PRDAADTDRI MNHSSILRKY KNNIMVVKDD INHPMSVVSS TKSRLALQHG DGHVVDYLSQ PVIDYILKSQ LYINASG
NMNAT2 Protein
100 μg
< 1.0 EU per ug protein as determined by the LAL method.
Specific activity: > 500 pmol/min/ug. One unit will convert 1.0 pmole of beta-NADH per minute to beta-NAD at PH 8.0 at 37C.
Human
His
NMNAT2 protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 20% glycerol and 1mM DTT.
Videos
Documentos