missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NFKBID Protein
A cDNA sequence encoding the NFKBID was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12837-5ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NFKBID Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMEAGPWR VSAPPSGPPQ FPAVVPGPSL EVARAHMLAL GPQQLLAQDE EGDTLLHLFA ARGLRWAAYA AAEVLQVYRR LDIREHKGKT PLLVAAAANQ PLIVEDLLNL GAEPNAADHQ GRSVLHVAAT YGLPGVLLAV LNSGVQVDLE ARDFEGLTPL HTAILALNVA MRPSDLCPRV LSTQARDRLD CVHMLLQMGA NHTSQEIKSN KTVLHLAVQA ANPTLVQLLL ELPRGDLRTF VNMKAHGNTA LHMAAALPPG PAQEAIVRHL LAAGADPTLR NLENEQPVHL LRPGPGPEGL RQLLKRSRVA PPGLSS | |
Greater than 95.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The NFKBID solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 10% glycerol and 1mM DTT. |