missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Neutrophil defensin 1 Protein

A DNA sequence encoding the Homo sapiens (Human) Neutrophil defensin 1, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP5924-ec-50ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15928719

  • 352.00€ / 50 microgramos


Fecha estimada de envío
29-06-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_001035965.1
Neutrophil defensin 1 Protein
50 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
1667
Greater than 80% as determined by SDS-PAGE.
Research Use Only
DEFA1
Recombinant Protein
E. coli
DIPEVVVSLA WDESLAPKHP GSRKNMACYC RIPACIAGER RYGTCIYQGR LWAFCC
Videos
Documentos