missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Neuronal calcium sensor 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Neuronal calcium sensor 1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6056-ec-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001122298.1 | |
Neuronal calcium sensor 1 Protein | |
Research Use Only | |
NCS1 | |
Recombinant Protein | |
E. coli | |
GKSNSKLKPE VVEELTRKTY FTEKEVQQWY KGFIKDCPSG QLDAAGFQKI YKQFFPFGDP TKFATFVFNV FDENKDGRIE FSEFIQALSV TSRGTLDEKL RWAFKLYDLD NDGYITRNEM LDIVDAIYQM VGNTVELPEE ENTPEKRVDR IFAMMDKNAD GKLTLQEFQE GSKADPSIVQ ALSLYDGLV | |
Greater than 80% as determined by SDS-PAGE. |
23413 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |