missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NEIL1 Protein
A DNA sequence encoding the Homo sapiens (Human) NEIL1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7940-ec-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001243481.1 | |
NEIL1 Protein | |
Research Use Only | |
NEIL1 | |
Recombinant Protein | |
E. coli | |
PEGPELHLAS QFVNEACRAL VFGGCVEKSS VSRNPEVPFE SSAYRISASA RGKELRLILS PLPGAQPQQE PLALVFRFGM SGSFQLVPRE ELPRHAHLRF YTAPPGPRLA LCFVDIRRFG RWDLGGKWQP GRGPCVLQEY QQFRENVLRN LADKAFDRPI CEALLDQRFF NGIGNYLRAE ILYRLKIPPF EKARSVLEAL QQHRPSPELT LSQKIRTKLQ NPDLLELCHS VPKEVVQLGG KGYGSESGEE DFAAFRAWLR CYGMPGMSSL QDRHGRTIWF QGDPGPLAPK GRKSRKKKSK ATQLSPEDRV EDALPPSKAP SRTRRAKRDL PKRTATQRPE GTSLQQDPEA PTVPKKGRRK GRQAASGHCR PRKVKADIPS LEPEGTSAS | |
Greater than 90% as determined by SDS-PAGE. |
79661 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |