Learn More
enQuireBio™ Recombinant Human NAIF1 Protein
A cDNA sequence encoding the NAIF1 was constructed and used to recombinantly synthesize the protein.
Marca:  enQuireBio™ QP12786-100ug
Detalles adicionales : Peso : 0.01000kg
 Ver más versiones de este producto
Código de producto. 15911539
  • 1815.00€ / 100 microgramos
Fecha estimada de envío: 29-05-2024
para ver el stock
Especificaciones
203245
100 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMAVPAKK RKMNFSEREV EIIVEELELK KHLLVNHFNA GVPLAAKSAA WHGILRRVNA VATCRRELPE VKKKWSDLKT EVRRKVAQVR AAVEGGEAPG PTEEDGAGGP GTGGGSGGGG PAVAPVLLTP MQQRICNLLG EATIISLPST TEIHPVALGP SATAAAATVT LTQIPTETTY HTLEEGVVEY CTAEAPPPLP PETPVDMMAQ HADTSVKPQA LKSRIALNSA KLIQEQRVTN LHVKEIAQHL EQQNDLLQMI RRSQEVQACA QERQAQAMEG TQAALSVLIQ VLRPMIKDFR RYLQSNTANP APASDPGQVA QNGQPDSIIQ
Greater than 90.0% as determined by SDS-PAGE.
NAIF1 Protein
Research Use Only
NAIF1
Human
His
NAIF1 protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 30% glycerol and 1mM DTT.