missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Myosin light chain 3 Protein

A DNA sequence encoding the Homo sapiens (Human) Myosin light chain 3, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP6401-ec-10ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16053050

  • 276.00€ / 10 microgramos


Fecha estimada de envío
20-09-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_000249.1
Myosin light chain 3 Protein
10 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
4634
Greater than 80% as determined by SDS-PAGE.
Research Use Only
MYL3
Recombinant Protein
E. coli
APKKPEPKKD DAKAAPKAAP APAPPPEPER PKEVEFDASK IKIEFTPEQI EEFKEAFMLF DRTPKCEMKI TYGQCGDVLR ALGQNPTQAE VLRVLGKPRQ EELNTKMMDF ETFLPMLQHI SKNKDTGTYE DFVEGLRVFD KEGNGTVMGA ELRHVLATLG ERLTEDEVEK LMAGQEDSNG CINYEAFVKH IMSS
Videos
Documentos