missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Myosin-9 Protein
A DNA sequence encoding the Homo sapiens (Human) Myosin-9, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8366-ec-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_002464.1 | |
Myosin-9 Protein | |
Research Use Only | |
MYH9 | |
Recombinant Protein | |
E. coli | |
AQQAADKYLY VDKNFINNPL AQADWAAKKL VWVPSDKSGF EPASLKEEVG EEAIVELVEN GKKVKVNKDD IQKMNPPKFS KVEDMAELTC LNEASVLHNL KERYYSGLIY TYSGLFCVVI NPYKNLPIYS EEIVEMYKGK KRHEMPPHIY AITDTAYRSM MQDREDQSIL CTGESGAGKT ENTKKVIQYL AYVASSHKSK KDQGELERQL LQANPILEAF GNAKTVKNDN SSRFGKFIRI | |
Greater than 80% as determined by SDS-PAGE. |
4627 | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |