missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MYLPF Protein
A DNA sequence encoding the Homo sapiens (Human) MYLPF, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8002-ec-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001311387.1 | |
MYLPF Protein | |
Research Use Only | |
MYLPF | |
Recombinant Protein | |
E. coli | |
APKRAKRRTV EGGSSSVFSM FDQTQIQEFK EAFTVIDQNR DGIIDKEDLR DTFAAMGRLN VKNEELDAMM KEASGPINFT VFLTMFGEKL KGADPEDVIT GAFKVLDPEG KGTIKKKFLE ELLTTQCDRF SQEEIKNMWA AFPPDVGGNV DYKNICYVIT HGDAKDQE | |
Greater than 90% as determined by SDS-PAGE. |
29895 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |